Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gp1bb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001999.1 | NP_001001999.1 | 1 | 1883 | ||
NM_010327.2 | NP_034457.1 | 2 | 1645 | |||
GenBank mRNA | AA222119.1 | - | 1 | 332 | ||
BC053108.1 | - | 1 | 1883 | |||
BC138156.1 | - | 1 | 1837 | 8861175 | ||
BC145160.1 | - | 1 | 1837 | 9053711 |