Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpaa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010331.2 | NP_034461.1 | 4 | 536 | ||
GenBank mRNA | AB002136.1 | - | 4 | 465 | ||
AB006970.1 | - | 4 | 472 | |||
AK077518.1 | - | 4 | 536 | |||
AK164219.1 | - | 4 | 482 | |||
AK164978.1 | - | 4 | 536 | |||
AK168057.1 | - | 4 | 739 | |||
AK192435.1 | - | 1 | 33 | |||
BC006697.1 | - | 4 | 475 | 3498573 |