Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002799.3 | NP_002790.1 | 3 | 288 | ||
GenBank mRNA | AK098757.1 | - | 3 | 257 | ||
AK223007.1 | - | 3 | 241 | |||
AK223356.1 | - | 3 | 253 | |||
AK303460.1 | - | 3 | 257 | |||
AK308756.1 | - | 3 | 288 | |||
AK314302.1 | - | 3 | 257 | |||
BC000509.2 | - | 3 | 244 | 2822305 | ||
BC008414.1 | - | 3 | 254 | |||
BC017116.2 | - | 2 | 179 | |||
BT007218.1 | - | 3 | 240 | |||
CR456721.1 | - | 3 | 240 | |||
D38048.1 | - | 3 | 254 | |||
GU727627.1 | - | 3 | 257 | |||
U44425.1 | - | 3 | 228 |