Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ONECUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004498.2 | NP_004489.1 | 2 | 1373 | ||
NR_073510.1 | 2 | 429 | ||||
GenBank mRNA | BC140830.1 | - | 2 | 1340 | 9021698 | |
BP289459.1 | - | 2 | 190 | |||
BX118406.1 | - | 2 | 429 | |||
EU532019.1 | - | 2 | 993 | |||
U77975.1 | - | 2 | 726 | |||
U96173.1 | - | 2 | 1373 |