Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181796.2 | NP_861461.1 | 5, 6 | 388 | ||
GenBank mRNA | BC064781.1 | - | 5, 6 | 388 | 30112237 | |
BC094623.1 | - | 5, 6 | 392 | 1381023 | ||
BC100587.1 | - | 5, 6 | 404 | |||
GQ181117.1 | - | 3, 4 | 298 |