Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252555.1 | NP_001239484.1 | 4 | 538 | ||
NM_010363.4 | NP_034493.1 | 3 | 235 | |||
GenBank mRNA | AB041613.1 | - | 4 | 538 | ||
AF093418.1 | - | 3 | 184 | |||
AK002398.1 | - | 3 | 192 | |||
AK012796.1 | - | 3 | 235 | |||
AK075927.1 | - | 3 | 189 | |||
BC031777.1 | - | 3 | 224 | 4168439 | ||
BY072664.1 | - | 3 | 236 |