Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HNRNPC SIRNA Assays ›
Gene Symbol:
HNRNPC
Gene Name:
heterogeneous nuclear ribonucleoprotein C (C1/C2)
Gene Aliases:
C1, C2, HNRNP, HNRPC, SNRPC
Chromosome Location:
Chr.14: 21209136 - 21269479 on Build GRCh38
UniGene ID:
Hs.508848
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s6719
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HNRNPC
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077442.1
NP_001070910.1
4
611
NM_001077443.1
NP_001070911.1
4
572
NM_004500.3
NP_004491.2
5
598
NM_031314.2
NP_112604.2
5
637
XM_006720125.2
XP_006720188.1
5
1254
XM_011536708.1
XP_011535010.1
6
671
XM_011536709.2
XP_011535011.1
5
1092
XM_011536710.2
XP_011535012.1
5
1293
XM_011536711.2
XP_011535013.1
5
1300
XM_011536712.1
XP_011535014.1
6
632
XM_017021252.1
XP_016876741.1
5
1261
XM_017021253.1
XP_016876742.1
5
1052
GenBank mRNA
AB209527.1
-
5
571
AK126950.1
-
4
3306
AK223517.1
-
5
576
AK225032.1
-
5
561
AK292619.1
-
7
914
AK298906.1
-
4
269
AK302213.1
-
4
535
AK312338.1
-
5
543
AK312382.1
-
4
557
BC003394.1
-
5
529
3447724
BC007052.1
-
4
557
3686841
BC008364.1
-
5
536
3354131
BC008423.1
-
4
557
4102910
BC066932.1
-
4
517
4375450
BC089438.1
-
4
493
30555817
BC103758.1
-
4
517
6187512
BC108658.1
-
4
462
6501821
BX161480.1
-
4
467
BX247961.1
-
4
328
BX247992.1
-
4
483
M16342.1
-
5
476
M29063.1
-
5
572
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splicing, via spliceosome
osteoblast differentiation
RNA splicing
gene expression
protein sumoylation
negative regulation of telomere maintenance via telomerase
ATP-dependent chromatin remodeling
3'-UTR-mediated mRNA stabilization
nucleotide binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
RNA binding
mRNA 3'-UTR binding
protein binding
poly(U) RNA binding
nucleosomal DNA binding
identical protein binding
poly(A) RNA binding
telomerase RNA binding
N6-methyladenosine-containing RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0