Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfdn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185182.1 | NP_001172111.1 | 3 | 160 | ||
NM_010385.2 | NP_034515.1 | 2 | 364 | |||
GenBank mRNA | AA023721.1 | - | 2 | 140 | ||
AK010679.1 | - | 2 | 354 | |||
AK076015.1 | - | 2 | 347 | |||
AK167900.1 | - | 2 | 350 | |||
AK177303.1 | - | 2 | 232 | |||
AV446044.1 | - | 1 | 19 | |||
BC022974.1 | - | 2 | 330 | 5373335 | ||
BY238879.1 | - | 2 | 364 | |||
KJ650212.1 | - | 2 | 108 | |||
M65255.1 | - | 2 | 327 |