Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hbb-b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016956.3 | NP_058652.1 | 2 | 223 | ||
GenBank mRNA | AB020014.1 | - | 2 | 166 | ||
AB020016.1 | - | 2 | 166 | |||
AB020017.1 | - | 2 | 166 | |||
AB364478.1 | - | 2 | 169 | |||
AF149782.1 | - | 2 | 169 | |||
BC027434.1 | - | 2 | 203 | 4920093 | ||
BC032264.1 | - | 2 | 214 | 3988455 |