Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hexa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010421.4 | NP_034551.2 | 8 | 994 | ||
GenBank mRNA | AK075895.1 | - | 8 | 970 | ||
AK075911.1 | - | 8 | 995 | |||
AK080777.1 | - | 8 | 967 | |||
AK144168.1 | - | 8 | 1122 | |||
AK150444.1 | - | 8 | 965 | |||
AK151960.1 | - | 8 | 969 | |||
AK159091.1 | - | 9 | 965 | |||
AK159814.1 | - | 8 | 980 | |||
AK159890.1 | - | 8 | 967 | |||
AK160112.1 | - | 8 | 954 | |||
AK167013.1 | - | 8 | 980 | |||
AK168186.1 | - | 8 | 994 | |||
AK170903.1 | - | 8 | 981 | |||
AK218113.1 | - | 3 | 341 | |||
BC010755.1 | - | 8 | 963 | 4225233 | ||
U07631.1 | - | 8 | 986 | |||
X64331.1 | - | 8 | 937 |