Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hgd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013547.3 | NP_038575.2 | 10 | 902 | ||
GenBank mRNA | AK002584.1 | - | 9 | 775 | ||
BC024369.1 | - | 9 | 716 | 4974534 | ||
BC037628.1 | - | 10 | 816 | 4973560 | ||
BC044758.2 | - | 9 | 702 | 5099956 | ||
BC055029.1 | - | 10 | 783 | 5054479 |