Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmg20b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163165.1 | NP_001156637.1 | 4 | 365 | ||
NM_001163166.1 | NP_001156638.1 | 4 | 262 | |||
NM_010440.3 | NP_034570.1 | 3 | 449 | |||
GenBank mRNA | AF146224.1 | - | 3 | 432 | ||
AK153816.1 | - | 4 | 263 | |||
AK155615.1 | - | 4 | 366 | |||
AK155652.1 | - | 4 | 267 | |||
AK155890.1 | - | 3 | 325 | |||
AK192784.1 | - | 2 | 135 | |||
AK192832.1 | - | 2 | 135 | |||
AK207354.1 | - | 2 | 136 | |||
AL355734.1 | - | 3 | 444 | |||
BC003924.1 | - | 3 | 337 | 3485302 | ||
BC011334.1 | - | 4 | 298 | 4012515 | ||
BC071217.1 | - | 1 | 1326 |