Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030661.4 | NP_109377.1 | 3 | 2130 | ||
NM_153631.2 | NP_705895.1 | 4 | 2269 | |||
XM_005249730.2 | XP_005249787.1 | 6 | 2717 | |||
XM_005249731.2 | XP_005249788.1 | 3 | 2271 | |||
XM_005249732.3 | XP_005249789.1 | 3 | 2136 | |||
XM_006715715.2 | XP_006715778.1 | 7 | 2833 | |||
XM_011515343.2 | XP_011513645.1 | 7 | 5981 | |||
GenBank mRNA | AK056567.1 | - | 4 | 2273 | ||
AK094200.1 | - | 3 | 1645 | |||
BC015180.1 | - | 3 | 2127 | 3905733 |