Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136066.2 | NP_001129538.1 | 4 | 760 | ||
NM_010443.2 | NP_034573.2 | 4 | 776 | |||
GenBank mRNA | AF029874.1 | - | 4 | 773 | ||
AF054669.1 | - | 4 | 777 | |||
AF054670.1 | - | 4 | 711 | |||
AK032175.1 | - | 4 | 718 | |||
AK134536.1 | - | 4 | 735 | |||
AK141426.1 | - | 4 | 754 | |||
AK152936.1 | - | 4 | 699 | |||
BC002011.1 | - | 4 | 732 | 3484259 |