Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170981.1 | NP_001164452.1 | 3 | 367 | ||
NM_001170982.1 | NP_001164453.1 | 2 | 341 | |||
NM_001170983.1 | NP_001164454.1 | 3 | 367 | |||
NM_001170984.1 | NP_001164455.1 | 3 | 367 | |||
NM_016884.3 | NP_058580.1 | 3 | 367 | |||
GenBank mRNA | AF095257.1 | - | 3 | 321 | ||
AK010845.1 | - | 3 | 346 | |||
AK011336.1 | - | 3 | 339 | |||
AK012633.2 | - | 3 | 329 | |||
AK019958.1 | - | 3 | 1542 | |||
AK088245.1 | - | 3 | 344 | |||
AK088678.1 | - | 3 | 339 | |||
AK089061.1 | - | 3 | 347 | |||
AK150399.1 | - | 3 | 343 | |||
AK151446.1 | - | 3 | 344 | |||
AK153053.1 | - | 3 | 344 | |||
AK166590.1 | - | 3 | 345 | |||
AK168802.1 | - | 3 | 341 | |||
AK169287.1 | - | 3 | 339 | |||
AK212671.1 | - | 1 | 193 | |||
AK214960.1 | - | 2 | 229 | |||
BC004706.1 | - | 3 | 329 | 3499283 | ||
BC095922.1 | - | 2 | 282 | 4988965 | ||
CK793036.1 | - | 321 | ||||
DV652725.1 | - | 3 | 391 |