Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxa10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008263.3 | NP_032289.2 | 1 | 504 | ||
GenBank mRNA | BC050839.1 | - | 1 | 476 | 6511608 | |
L08757.1 | - | 1 | 504 |