Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010455.2 | NP_034585.1 | 2 | 1730 | ||
GenBank mRNA | AK033500.1 | - | 2 | 632 | ||
BC036986.1 | - | 4 | 697 | |||
BC131978.1 | - | 2 | 594 | 40130255 | ||
BC132643.1 | - | 2 | 594 | 40130920 | ||
M17192.1 | - | 2 | 662 |