Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175730.5 | NP_783857.1 | 2 | 549 | ||
GenBank mRNA | AK078527.1 | - | 2 | 550 | ||
BC115551.1 | - | 2 | 550 | 40101712 | ||
BC115552.1 | - | 2 | 550 | 40101713 |