Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290730.1 | NP_001277659.1 | 1 | 458 | ||
NM_008276.3 | NP_032302.2 | 1 | 458 | |||
GenBank mRNA | AK015019.1 | - | 1 | 162 |