Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hpx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017371.2 | NP_059067.2 | 2, 3 | 173 | ||
GenBank mRNA | AK145928.1 | - | 2, 3 | 171 | ||
BC011246.1 | - | 2, 3 | 153 | 4196684 | ||
BC019901.1 | - | 2, 3 | 155 | 5123914 | ||
BY088352.1 | - | 2, 3 | 174 | |||
U89889.1 | - | 2, 3 | 129 |