Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iapp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010491.2 | NP_034621.1 | 3 | 543 | ||
GenBank mRNA | BC027527.1 | - | 3 | 514 | 1379049 | |
M25389.1 | - | 2 | 450 |