Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Id2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010496.3 | NP_034626.1 | 1 | 190 | ||
GenBank mRNA | AK003222.1 | - | 1 | 187 | ||
AK013239.1 | - | 1 | 185 | |||
AK013378.1 | - | 1 | 188 | |||
AK135547.1 | - | 1 | 192 | |||
AK146346.1 | - | 1 | 190 | |||
AK146606.1 | - | 1 | 193 | |||
BC006921.1 | - | 1 | 167 | 2655173 | ||
BC053699.1 | - | 1 | 177 | 6515664 | ||
M69293.1 | - | 1 | 184 |