Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifrd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025903.2 | NP_080179.1 | 11 | 1337 | ||
GenBank mRNA | AK002702.1 | - | 5 | 562 | ||
AK007687.1 | - | 11 | 1338 | |||
AK149496.1 | - | 11 | 1318 | |||
AK156625.1 | - | 10 | 1413 | |||
AK165967.1 | - | 11 | 1331 | |||
AK166130.1 | - | 11 | 1405 | |||
AK166258.1 | - | 11 | 1295 | |||
AY298860.1 | - | 11 | 1166 | |||
BC023486.1 | - | 11 | 1284 | |||
BC037434.1 | - | 11 | 1293 | 2651172 |