Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igfbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008342.3 | NP_032368.2 | 4 | 866 | ||
GenBank mRNA | AK011784.1 | - | 5 | 736 | ||
AK011790.1 | - | 4 | 795 | |||
AK012703.1 | - | 4 | 858 | |||
BC012724.1 | - | 4 | 854 | 2654907 | ||
BC054473.1 | - | 4 | 839 | 6488878 | ||
DV552718.1 | - | 3 | 394 | |||
X81580.1 | - | 4 | 837 |