Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Incenp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016692.3 | NP_057901.2 | 9 | 1504 | ||
GenBank mRNA | AB100432.1 | - | 8 | 1298 | ||
AB100433.1 | - | 8 | 1298 | |||
AF117610.1 | - | 9 | 1440 | |||
AK034764.1 | - | 9 | 1433 | |||
AK081841.1 | - | 9 | 1438 | |||
AK088627.1 | - | 9 | 1461 | |||
BC052414.1 | - | 9 | 1437 | 6401188 |