Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Invs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281977.1 | NP_001268906.1 | 10 | 1592 | ||
NM_001281978.1 | NP_001268907.1 | 12 | 1870 | |||
NM_010569.4 | NP_034699.3 | 10 | 1644 | |||
GenBank mRNA | AF034860.3 | - | 10 | 1586 | ||
AK041671.1 | - | 9 | 1411 | |||
AK158766.1 | - | 12 | 1829 | |||
BC126929.1 | - | 10 | 1481 | 40081431 |