Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008404.4 | NP_032430.2 | 14 | 2131 | ||
GenBank mRNA | AK088435.1 | - | 14 | 2120 | ||
AK136502.1 | - | 14 | 2132 | |||
AK161236.1 | - | 13 | 1855 | |||
AK169587.1 | - | 12 | 1733 | |||
BC145644.1 | - | 13 | 2053 | 40130947 | ||
M31039.1 | - | 13 | 2019 | |||
X14951.1 | - | 14 | 2097 |