Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159564.1 | NP_001153036.1 | 14 | 2056 | ||
NM_021359.3 | NP_067334.1 | 11 | 1950 | |||
GenBank mRNA | AF115376.1 | - | 14 | 1902 | ||
AK036439.1 | - | 14 | 2057 | |||
AK085900.1 | - | 14 | 2097 | |||
BC049185.1 | - | 11 | 1919 | 6475423 |