Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cuzd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008411.3 | NP_032437.3 | 8 | 1819 | ||
GenBank mRNA | BC140992.1 | - | 8 | 1754 | 9055604 | |
U69699.2 | - | 8 | 1818 |