Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itpr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010585.5 | NP_034715.3 | 43 | 5575 | ||
GenBank mRNA | CB521690.1 | - | 6 | 427 | ||
CF168791.1 | - | 5 | 497 | |||
S75867.1 | - | 4 | 206 | |||
X15373.1 | - | 43 | 5553 |