Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048177.2 | NP_001041642.1 | 9 | 1610 | ||
NM_008413.3 | NP_032439.2 | 10 | 1693 | |||
GenBank mRNA | AK087255.1 | - | 9 | 1577 | ||
BC054807.1 | - | 9 | 1373 | 6838318 | ||
BC059834.1 | - | 9 | 1373 | 6838318 | ||
L16956.1 | - | 9 | 1362 |