Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcna5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145983.2 | NP_666095.1 | 1 | 1421 | ||
GenBank mRNA | AF108659.1 | - | 1 | 1278 | ||
AK146843.1 | - | 1 | 1238 | |||
BC021787.1 | - | 1 | 1219 | 4527103 | ||
L22218.1 | - | 1 | 1326 |