Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008420.4 | NP_032446.2 | 2 | 1324 | ||
GenBank mRNA | BC031776.1 | - | 2 | 1280 | 4507847 | |
BC051422.1 | - | 2 | 1281 | 6493266 | ||
BC061501.1 | - | 2 | 1305 | 4506061 | ||
M64228.1 | - | 2 | 1129 |