Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021542.4 | NP_067517.1 | 4 | 891 | ||
GenBank mRNA | AF259395.1 | - | 4 | 561 | ||
AF319542.1 | - | 4 | 630 | |||
AK085330.1 | - | 4 | 892 | |||
AK132172.1 | - | 4 | 919 | |||
AK172608.1 | - | 4 | 892 | |||
BC034012.1 | - | 4 | 893 | 4239113 | ||
BC051088.1 | - | 4 | 860 | |||
BC058164.1 | - | 4 | 882 | 5323480 |