Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163510.1 | NP_001156982.1 | 1 | 298 | ||
NM_008433.4 | NP_032459.3 | 1 | 298 | |||
GenBank mRNA | AF042487.1 | - | 1 | 164 | ||
AF072884.1 | - | 1 | 124 | |||
AK010943.1 | - | 1 | 299 | |||
AK085752.1 | - | 1 | 108 | |||
AK155910.1 | - | 1 | 263 | |||
BC010274.1 | - | 1 | 247 | 3589411 | ||
BY197732.1 | - | 1 | 297 |