Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163510.1 | NP_001156982.1 | 4 | 901 | ||
NM_008433.4 | NP_032459.3 | 4 | 901 | |||
GenBank mRNA | AB128930.1 | - | 2 | 664 | ||
AF042487.1 | - | 4 | 767 | |||
AF072884.1 | - | 4 | 727 | |||
AK010943.1 | - | 4 | 902 | |||
AK085752.1 | - | 4 | 718 | |||
AK155910.1 | - | 4 | 866 | |||
BC010274.1 | - | 4 | 850 | 3589411 |