Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010615.1 | NP_034745.1 | 9, 10 | 1313 | ||
GenBank mRNA | AJ223293.1 | - | 8, 9 | 1008 | ||
AK083324.1 | - | 9, 10 | 1290 | |||
AK147754.1 | - | 9, 10 | 1337 | |||
BC060670.1 | - | 9, 10 | 1313 | 6406754 |