Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPCAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258357.1 | NP_001245286.1 | 5 | 1213 | ||
NM_001258358.1 | NP_001245287.1 | 5 | 1225 | |||
NM_001258359.1 | NP_001245288.1 | 5 | 1619 | |||
NM_002149.3 | NP_002140.2 | 5 | 1439 | |||
NM_134421.2 | NP_602293.1 | 6 | 1581 | |||
XM_005246160.1 | XP_005246217.1 | 6 | 1504 | |||
XM_005246161.1 | XP_005246218.1 | 6 | 1627 | |||
XM_005246162.1 | XP_005246219.1 | 6 | 1630 | |||
XM_005246163.1 | XP_005246220.1 | 7 | 1677 | |||
XM_011510346.2 | XP_011508648.1 | 5 | 1283 | |||
XM_011510347.1 | XP_011508649.1 | 7 | 1724 | |||
XM_011510348.1 | XP_011508650.1 | 6 | 1517 | |||
XM_011510349.1 | XP_011508651.1 | 6 | 1543 | |||
XM_017003950.1 | XP_016859439.1 | 7 | 3171 | |||
XM_017003951.1 | XP_016859440.1 | 7 | 1654 | |||
XM_017003952.1 | XP_016859441.1 | 5 | 1399 | |||
XM_017003953.1 | XP_016859442.1 | 6 | 1534 | |||
GenBank mRNA | AF070616.1 | - | 3 | 808 | ||
AK000596.1 | - | 6 | 1587 | |||
AK093973.1 | - | 1 | 1944 | |||
BC009846.2 | - | 5 | 1414 | 4100091 | ||
BC017028.1 | - | 5 | 1406 | 3902811 | ||
BC017482.1 | - | 5 | 1429 | 3504942 | ||
CR749837.1 | - | 8 | 4723 |