Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kifc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010630.2 | NP_034760.2 | 11 | 1236 | ||
GenBank mRNA | AK144304.1 | - | 12 | 1395 | ||
D49545.1 | - | 11 | 1312 | |||
U92949.1 | - | 11 | 1226 |