Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klra3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289604.1 | NP_001276533.1 | 4 | 518 | ||
NM_001289605.1 | NP_001276534.1 | 4 | 552 | |||
NM_010648.3 | NP_034778.2 | 4 | 518 | |||
GenBank mRNA | AK159595.1 | - | 4 | 467 | ||
BC099867.1 | - | 350 | 40040425 | |||
BC099878.1 | - | 350 | 40040424 | |||
BC099879.1 | - | 350 | 40040423 | |||
BC099880.1 | - | 350 | 40040422 | |||
BC101952.1 | - | 4 | 394 | 40044699 | ||
BC103543.1 | - | 4 | 394 | 40044703 | ||
BC103544.1 | - | 4 | 428 | 40044701 | ||
BC103545.1 | - | 4 | 394 | 40044700 | ||
BC132524.1 | - | 4 | 399 | 40130801 | ||
BC132526.1 | - | 4 | 399 | 40130803 | ||
BC145192.1 | - | 4 | 399 | 9053743 | ||
BC145193.1 | - | 4 | 433 | 9053744 | ||
U09739.1 | - | 4 | 518 | |||
U10305.1 | - | 4 | 434 | |||
U49868.1 | - | 4 | 499 | |||
U56404.1 | - | 4 | 393 | |||
U56405.1 | - | 4 | 393 |