Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008471.2 | NP_032497.1 | 2 | 520 | ||
GenBank mRNA | AB472075.1 | - | 2 | 520 | ||
BC034561.1 | - | 2 | 513 | 2655016 | ||
BC085304.1 | - | 2 | 520 | 6437375 | ||
M28698.1 | - | 2 | 515 |