Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010669.2 | NP_034799.2 | 9 | 2054 | ||
GenBank mRNA | AK009241.1 | - | 6 | 1160 | ||
AK137688.1 | - | 9 | 2053 | |||
DV553143.1 | - | 1 | 120 |