Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGFG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006076.4 | NP_006067.3 | 2 | 398 | ||
XM_005250306.2 | XP_005250363.1 | 2 | 384 | |||
XM_017012112.1 | XP_016867601.1 | 2 | 339 | |||
XM_017012113.1 | XP_016867602.1 | 2 | 384 | |||
XM_017012114.1 | XP_016867603.1 | 2 | 384 | |||
GenBank mRNA | AF015042.1 | - | 2 | 322 | ||
AK314954.1 | - | 2 | 384 | |||
BC009393.2 | - | 2 | 397 | |||
BC017329.2 | - | 2 | 350 | 4810320 | ||
BF310674.1 | - | 2 | 395 | |||
BF314987.1 | - | 2 | 398 |