Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133664.3 | NP_598425.2 | 3 | 1102 | ||
GenBank mRNA | AK083526.1 | - | 3 | 1103 | ||
BC016257.1 | - | 3 | 773 | 4912735 | ||
BC025580.1 | - | 3 | 1006 | 5325039 | ||
BC031131.1 | - | 3 | 949 | 4954891 |