Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008483.3 | NP_032509.2 | 9 | 1106 | ||
GenBank mRNA | AK140351.1 | - | 9 | 1085 | ||
AK158444.1 | - | 9 | 1078 | |||
AK194224.1 | - | 2 | 185 | |||
AK205168.1 | - | 1 | 36 | |||
BC026051.1 | - | 9 | 1033 | 4036357 |