Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Laptm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010686.3 | NP_034816.1 | 4 | 292 | ||
GenBank mRNA | AF364050.1 | - | 4 | 310 | ||
AK031802.1 | - | 4 | 306 | |||
AK050633.1 | - | 4 | 307 | |||
AK089432.1 | - | 4 | 342 | |||
AK089508.1 | - | 4 | 315 | |||
AK089535.1 | - | 4 | 307 | |||
AK150299.1 | - | 4 | 303 | |||
AK152918.1 | - | 4 | 303 | |||
AK153877.1 | - | 4 | 306 | |||
AK155744.1 | - | 4 | 300 | |||
AK163146.1 | - | 4 | 307 | |||
AK170438.1 | - | 4 | 294 | |||
AK170690.1 | - | 4 | 326 | |||
AK170867.1 | - | 4 | 310 | |||
AK171053.1 | - | 4 | 317 | |||
AK171057.1 | - | 4 | 342 | |||
AK171105.1 | - | 4 | 307 | |||
AK171116.1 | - | 4 | 307 | |||
AK171149.1 | - | 4 | 315 | |||
AK171172.1 | - | 307 | ||||
AK171193.1 | - | 4 | 307 | |||
AK171213.1 | - | 4 | 307 | |||
AK171236.1 | - | 4 | 307 | |||
AK171244.1 | - | 4 | 307 | |||
AK171250.1 | - | 4 | 328 | |||
AK171269.1 | - | 4 | 344 | |||
AK171270.1 | - | 4 | 310 | |||
AK171274.1 | - | 4 | 300 | |||
AK171280.1 | - | 4 | 340 | |||
AK171285.1 | - | 4 | 344 | |||
AK171481.1 | - | 4 | 343 | |||
AK171547.1 | - | 4 | 307 | |||
AK171562.1 | - | 4 | 298 | |||
AK171571.1 | - | 4 | 324 | |||
AK171575.1 | - | 4 | 342 | |||
AK172306.1 | - | 4 | 307 | |||
AK172486.1 | - | 4 | 342 | |||
AK172707.1 | - | 4 | 307 | |||
AK172885.1 | - | 4 | 282 | |||
BC020993.1 | - | 4 | 292 | 4037686 | ||
BC055057.1 | - | 4 | 279 | 6529760 | ||
BC114403.1 | - | 4 | 272 | |||
BC158032.1 | - | 4 | 488 | 9007335 | ||
U29539.1 | - | 4 | 310 | |||
U51239.1 | - | 4 | 331 |