Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162432.1 | NP_001155904.1 | 5, 6 | 468 | ||
NM_001162433.1 | NP_001155905.1 | 5, 6 | 510 | |||
NM_010693.3 | NP_034823.1 | 4, 5 | 565 | |||
GenBank mRNA | AK088001.1 | - | 4, 5 | 566 | ||
BC011474.1 | - | 5, 6 | 475 | 4191610 | ||
BQ563042.1 | - | 5, 6 | 459 | |||
CA577097.1 | - | 2, 3 | 201 | |||
M12056.1 | - | 4, 5 | 541 | |||
X03533.1 | - | 4, 5 | 571 |