Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113408.1 | NP_001106879.1 | 4 | 574 | ||
NM_010697.1 | NP_034827.1 | 4 | 759 | |||
GenBank mRNA | AB221569.1 | - | 3 | 116 | ||
AB250383.1 | - | 3 | 113 | |||
AF030333.1 | - | 4 | 277 | |||
AK193238.1 | - | 3 | 116 | |||
BC013624.1 | - | 4 | 399 | 3708169 | ||
CK790183.1 | - | 4 | 579 | |||
U69270.1 | - | 4 | 759 | |||
U70375.1 | - | 3 | 109 | |||
U89488.1 | - | 4 | 357 |