Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhx8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010713.2 | NP_034843.2 | 4 | 613 | ||
GenBank mRNA | AJ000338.1 | - | 4 | 713 | ||
AK160121.1 | - | 4 | 614 | |||
AK165733.1 | - | 4 | 607 | |||
BC125281.1 | - | 4 | 522 | 40129796 | ||
BC125283.1 | - | 4 | 522 | 40129798 | ||
BC144768.1 | - | 4 | 522 | 9053316 | ||
D49658.1 | - | 4 | 582 |