Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lnx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159577.1 | NP_001153049.1 | 10 | 2239 | ||
NM_001159578.1 | NP_001153050.1 | 11 | 2384 | |||
NM_001159579.1 | NP_001153051.1 | 9 | 2015 | |||
NM_001159580.1 | NP_001153052.1 | 9 | 1985 | |||
NM_010727.4 | NP_034857.3 | 9 | 2015 | |||
GenBank mRNA | AF034745.1 | - | 10 | 2239 | ||
AF034746.1 | - | 9 | 1943 | |||
AK044127.1 | - | 9 | 1968 | |||
AK138326.1 | - | 9 | 1986 | |||
AK161194.1 | - | 11 | 2370 | |||
BC139771.1 | - | 9 | 1943 | 40111506 | ||
KJ418422.1 | - | 9 | 1713 |